Would you like to log in again?
Elite-Q PLOT Phase Columns, PerkinElmer
Supplier: PerkinElmer
N9316360
10137-180EA 2206.54 USD InStock
10137-180
Elite-Q PLOT Phase Columns, PerkinElmer
Chromatography Columns
Elite-Q PLOT Phase Columns are for analysis of light gases and hydrocarbons.
  • Available in a range of inner diameter measurements
  • Column length is 30 meters
  • Suited for analysis of light cases and hydrocarbons
PLOT columns are the best choice for analysis of highly volatile compounds such as permanent gases, solvents, and volatile petrochemicals such as refinery gases.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty