Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Boron, Sigma-Aldrich®

Supplier: MilliporeSigma

1.12070.0010 1.12070.0010
EM1.12070.0010EA 107 USD InStock
EM1.12070.0010 CA1.12070.0010
Boron, Sigma-Aldrich®
Boron


Formula: B
MW: 10.81 g/mol
MDL Number: MFCD00134034
CAS Number: 7440-42-8
UN: 3178
ADR: 4.1,III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty