Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

D-Gluconic acid 50% in aqueous solution, Sigma-Aldrich®

Supplier: MilliporeSigma
Synonyms: Gluconic acid

M-Clarity™ quality level = MQ200

8.22057.9025 8.22057.9025
EM8.22057.9025EA 882 USD InStock
CA8.22057.9025 EM8.22057.9025
D-Gluconic acid 50% in aqueous solution, Sigma-Aldrich®
D-Gluconic acid


Formula: HOH₂C(CH(OH))₄COOH
MW: 196.16 g/mol
Density: 1.24 g/cm³ (20 °C)
MDL Number: MFCD00004240
CAS Number: 526-95-4
UN: 3265
ADR: 8,III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty