Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sulfuric acid ≥98% for nitrogen analysis, Supelco®

Supplier: MilliporeSigma
Danger

M-Clarity™ quality level = MQ300

1.00748.0500 1.00748.0500
EM1.00748.0500CS 649 USD InStock
CA1.00748.0500 EM1.00748.0500
Sulfuric acid ≥98% for nitrogen analysis, Supelco®
Sulfuric acid


Formula: H₂SO₄
MW: 98.08 g/mol
Boiling Pt: ∼335 °C (1013 hPa)
Melting Pt: 3 °C
Density: 1.84 g/cm³ (20 °C)
MDL Number: MFCD00064589
CAS Number: 7664-93-9
UN: 1830
ADR: 8,II
Order Now
Specification Test Results
Assay (alkalimetric) ≥ 97.5 %
Total nitrogen (N) ≤ 1 ppm
Residue on ignition ≤ 2 ppm

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty