Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

n-Nonadecane, Sigma-Aldrich®

Supplier: MilliporeSigma
Synonyms: N-nonadecane

M-Clarity™ quality level = MQ200

8.22110.0010 8.22110.0010
EM8.22110.0010EA 35 USD InStock
EM8.22110.0010 CA8.22110.0010
n-Nonadecane, Sigma-Aldrich®
n-Nonadecane


Formula: H₃C(CH₂)₁₇CH₃
MW: 268.53 g/mol
Boiling Pt: 329.7 °C (1013 hPa)
Melting Pt: 32.5 °C
Density: 0.7845 g/cm³ (20 °C)
MDL Number: MFCD00009012
CAS Number: 629-92-5
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty