Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

L(-)-Malic acid, Sigma-Aldrich®

Supplier: MilliporeSigma
Synonyms: L-(–)-Malic Acid(S)-(-)-Hydroxysuccinic acidL(-)-Hydroxy-1,4-butanedioic acid(S)-(-)-Hydroxy-1,4-butanedioic acid(S)-2-hydroxysuccinic acidL(-)-Hydroxysuccinic acidL(-)-Malic acid

M-Clarity™ quality level = MQ200

8.00384.0005 8.00384.0005
EM8.00384.0005EA 31 USD InStock
EM8.00384.0005 CA8.00384.0005
L(-)-Malic acid, Sigma-Aldrich®
L(-)-Malic acid


Formula: C₄H₆O₅
MW: 134.09 g/mol
Boiling Pt: 322 °C (1013 hPa)
Melting Pt: 101…104 °C
Density: 1.595 g/cm³ (20 °C)
MDL Number: MFCD00064213
CAS Number: 97-67-6
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty