Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ethylenediammonium dichloride, Sigma-Aldrich®

Supplier: MilliporeSigma
Synonyms: Ethylenediamine dihydrochloride

M-Clarity™ quality level = MQ200

8.00948.9050 8.00948.9050
EM8.00948.9050EA 4810 USD InStock
EM8.00948.9050 CA8.00948.9050
Ethylenediammonium dichloride, Sigma-Aldrich®
Ethylenediammonium dichloride


Formula: C₂H₁₀Cl₂N₂
MW: 133.02 g/mol
MDL Number: MFCD00012524
CAS Number: 333-18-6
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty