Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Methacrylic acid, Sigma-Aldrich®

Supplier: MilliporeSigma
Synonyms: 2-Methylpropenoic acid

M-Clarity™ quality level = MQ200

8.00578.9025 8.00578.9025
EM8.00578.9025EA 814 USD InStock
CA8.00578.9025 EM8.00578.9025
Methacrylic acid, Sigma-Aldrich®
Methacrylic acid


Formula: C₄H₆O₂
MW: 86.09 g/mol
Boiling Pt: 163 °C (1013 hPa)
Melting Pt: 15 °C
Density: 1.02 g/cm³ (20 °C)
Flash Pt: 68 °C
MDL Number: MFCD00002651
CAS Number: 79-41-4
UN: 2531
ADR: 8,II
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty