Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N,N,N',N'-Tetramethyl-p-phenylenediamine dihydrochloride, Sigma-Aldrich®

Supplier: MilliporeSigma
Synonyms: N,N,N',N'-Tetramethyl-1,4-phenylenediamine dihydrochlorideN1,N1,N4,N4-Tetramethylbenzene-1,4-diamine dihydrochlorideTMPD 2 HClN,N,N',N'-Tetramethyl-1,4-phenylenediammonium dichloride

M-Clarity™ quality level = MQ200

8.21102.0002 8.21102.0002
EM8.21102.0002EA 87 USD InStock
EM8.21102.0002 CA8.21102.0002
N,N,N',N'-Tetramethyl-p-phenylenediamine dihydrochloride, Sigma-Aldrich®
N,N,N',N'-Tetramethyl-p-phenylenediamine dihydrochloride


Formula: C₁₀H₁₈Cl₂N₂
MW: 237.17 g/mol
Melting Pt: 227…230 °C
MDL Number: MFCD00012482
CAS Number: 637-01-4
https://us-legacy.vwr.com/stibo/web/std.lang.all/61/62/2346162.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty