Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Lithium iodide, Sigma-Aldrich®

Supplier: MilliporeSigma

M-Clarity™ quality level = MQ200

8.18287.0002 8.18287.0002
EM8.18287.0002EA 135 USD InStock
CA8.18287.0002 EM8.18287.0002
Lithium iodide, Sigma-Aldrich®
Lithium iodide


Formula: ILi
MW: 133.85 g/mol
Boiling Pt: 1177 °C (1013 hPa)
Melting Pt: 450 °C
Density: 3.49 g/cm³ (20 °C)
MDL Number: MFCD00011092
CAS Number: 10377-51-2
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty