Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Aminoguanidinium hydrogen carbonate, Sigma-Aldrich®

Supplier: MilliporeSigma
Synonyms: 1-Aminoguanidinium hydrogen carbonateAminoguanidine hydrogen carbonate

M-Clarity™ quality level = MQ200

8.01417.9025 8.01417.9025
EM8.01417.9025EA 1191 USD InStock
EM8.01417.9025 CA8.01417.9025
Aminoguanidinium hydrogen carbonate, Sigma-Aldrich®
Aminoguanidinium hydrogen carbonate


Formula: C₂H₈N₄O₃
MW: 136.11 g/mol
MDL Number: MFCD00012949
CAS Number: 2582-30-1
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty