Would you like to log in again?
TO-14A Aromatics Mix, Restek
Supplier: Restek
34404 26348 34423 26349
75795-126EA 3130.96 USD InStock
75795-126 75795-224 75795-222 75795-172
TO-14A Aromatics Mix, Restek
Standards Environmental Analysis Standards
Environmental Air Sampling Gas Standards meet lab requirements for two separate sources of calibration standards.
  • High-quality
  • No rental or demurrage fees required
  • Fast delivery provides freshest standard possible
Our high-quality air sampling gas calibration standards are provided by Spectra/Linde and Scott/Air Liquide—meeting lab requirements for two separate sources of calibration standards. Each comes with a certificate of analysis and unique serial number. All cylinders are disposable and do not require rental or demurrage fees. Recertification of cylinders is available directly with our suppliers. All cylinders are drop-shipped from our suppliers to provide fast delivery and the "freshest" standard possible. Minimum 12-month stability on all cylinders.
Ordering information: TO-14A Aromatics Mix contains the following components: Benzene; Chlorobenzene; m-Dichlorobenzene; o-Dichlorobenzene; p-Dichlorobenzene; Ethyl benzene; Styrene; Toluene; 1,2,4-Trichlorobenzene; 1,2,4-Trimethylbenzene; 1,3,5-Trimethylbenzene; m-Xylene; o-Xylene; p-Xylene.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty