Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

TO-15 Subset 25 Component Mix, Restek

Supplier: Restek
34434 26358 26357
75794-984EA 2806.97 USD InStock
75794-984 75795-146 75795-060
TO-15 Subset 25 Component Mix, Restek
Standards Environmental Analysis Standards
Environmental air sampling gas standards meet lab requirements for two separate sources of calibration standards.

  • High-quality
  • No rental or demurrage fees required
  • Fast delivery provides freshest standard possible
  • One year shelf life
  • Blend tolerance: ±10%

TO-15 Subset 25 component mix contains the following components: acetone; allyl chloride; benzyl chloride; bromodichloromethane; bromoform; 1,3-butadiene; 2-butanone (MEK); carbon disulfide; cyclohexane; dibromochloromethane; trans-1,2-dichloroethene; 1,4-dioxane; ethyl acetate; 4-ethyltoluene; heptane; hexane; 2-hexanone (MBK); 4-methyl-2-pentanone; methyl tert-butyl ether (MTBE); 2-propanol; propylene; tetrahydrofuran; 2,2,4-trimethylpentane; vinyl acetate; vinyl bromide.

Pressure: 240 psig (17 bar).

Caution: *Stability of this compound cannot be guaranteed.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty