Would you like to log in again?
Natural Gas Standard #1, Restek
Supplier: Restek
34438
75795-212EA 746.9 USD InStock
75795-212
Natural Gas Standard #1, Restek
Standards Petroleum Industry Standards
Natural gas standards with 10 components are available in three mixes, from lean to rich.
  • Cylinder: 13.16 L at 200 psig
  • Mini-regulator designed specially for these standards is also available
Each of these natural gas standards has an extended list of C6+ components.
Ordering information: Natural Gas Standard #1 contains the following compounds: Nitrogen, 1.000 %; Carbon dioxide, 0.500 %; Methane UHP, 94.750 %; Ethane UHP, 2.000 %; Propane, 0.750 %; Isobutane, 0.300 %; N-Butane, 0.300 %; Isopentane, 0.150 %; N-Pentane, 0.150 %; Hexanes plus**, 0.100 %.
Available in three mixes, Standards #1 to #3, from lean to rich. Each has an extended list of C6+ components.
Packaging: Ideal Heating Value: Dry BTU/SCF @ 14.696psia & 60°F.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty