Would you like to log in again?
Refinery Gas Standard #1, Restek
Supplier: Restek
34441
75795-220EA 1154.3 USD InStock
75795-220
Refinery Gas Standard #1, Restek
Standards Petroleum Industry Standards
Refinery gas standards are available in three mixes with varying C5 unsaturates or extended C6+ components.
  • Cylinder: 5.2 L at 70 psig
  • Mini-regulator designed specially for these standards is also available
Refinery gas standard #1 features 21 components.
Ordering information: Refinery Gas Standard #1 contains the following compounds: Hydrogen, 40.750 %; Argon, 0.500 %; Nitrogen, 4.000 %; Carbon monoxide, 1.000 %; Carbon dioxide, 3.000 %; Methane, 8.500 %; Ethane, 6.000 %; Ethylene, 2.000 %; Propane, 7.000 %; Propylene, 3.000 %; Propadiene, 0.850 %; Isobutane, 6.000 %; N-Butane, 4.000 %; Isobutylene, 2.000 %;1,3 Butadiene, 3.000 %;cis-2-Butene, 2.000 %;trans-2-Butene, 2.000 %;1-Bbutene, 2.000 %;Isopentane, 1.000 %;N-Pentane, 1.000 %;N-Hexane, 0.500 %.
Available in three mixes with varying C5 unsaturates or extended C6+ components.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty