Would you like to log in again?
Beakers, Jacketed, Low Form, Chemglass
Supplier: Chemglass
CG-1107-12
89028-336EA 1472.37 USD InStock
89028-336
Beakers, Jacketed, Low Form, Chemglass
Beakers
These beakers have a larger opening at the top of the jacket and a shorter overall height.
  • Width to height ratio on all sizes is 2:1
  • With a flat bottom on both the inner and outer vessel
  • Hose connections have an O.D. of 10 mm at the largest serration
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty