Would you like to log in again?
Imject™ Bovine Serum Albumin, Thermo Scientific
Supplier: Thermo Scientific
Pierce
77171 77171
PI77171EA 121.92 USD InStock
PI77171 CAPI77171
Imject™ Bovine Serum Albumin, Thermo Scientific
Proteins and Peptides
Imject™ Bovine Serum Albumin (BSA) in MES Buffer is a carrier protein that enable simple preparation of effective immunogens with amine- or carboxyl-peptide antigens.
  • Purified BSA (Fraction V) in MES buffer
  • Ready for use in glutaraldehyde, NHS-ester or EDC crosslinking methods
BSA is a popular carrier protein for conjugation to haptens and other weak antigens to make them more immunogenic for the purpose of antibody production
BSA is sufficiently large to function as the primary immunogen or it can be used as an irrelevant protein carrier for antibody screening and immunoassays after using KLH as the carrier protein to generate the immune response against the hapten.
It also provides an independent hapten-carrier protein for microplate and other assay techniques required to screen antibodies produced using KLH or another more immunogenic carrier.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty