Would you like to log in again?
ActiveHOOK™ HyperCarrier™ Bovine Serum Albumin (BSA), Cationized (active), G-Biosciences
Supplier: G-Biosciences
ActiveHOOK™ ActiveHOOK™ HyperCarrier™
786-097 786-097
82022-456EA 635.5 USD InStock
CA82022-456 82022-456
ActiveHOOK™ HyperCarrier™ Bovine Serum Albumin (BSA), Cationized (active), G-Biosciences
Proteins and Peptides
ActiveHOOK™ HyperCarrier™ is a pre‐activated form of HyperCarrier™ that has been activated by sulfoSMCC and is ready for direct coupling with peptides through the cysteine residues. HyperCarrier™ is a cationized form of bovine serum albumin. Cationized proteins are taken into antigen presenting cells (APC) quicker and more efficiently than normal proteins.  The positive charges on the cationized protein give it greater affinity for the negatively charged cell surface of the APCs. The cationized protein is internalized via activated receptor mediated endocytosis as opposed to pinocytosis (Border et al. (1982) J.Clin. Invest. 69:451). ActiveHOOK™ HyperCarrier™ is supplied lyophilized in 10mg/vial size.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty