Would you like to log in again?
Holmium Oxide Glass Filter, PerkinElmer
Supplier: PerkinElmer
N1010545
10139-532EA 1418.85 USD InStock
10139-532
Holmium Oxide Glass Filter, PerkinElmer
Standards Color Reference and Spectroscopy Standards
Holmium oxide glass filter for checking the wavelength accuracy of a UV/Vis or UV/Vis/NIR spectrometer.
  • Glass construction
  • Mounted on aluminum spring mount
  • Provided in a robust aluminum container
The filter is mounted in precision aluminum spring mount and is provided in a robust aluminum container, with a Certificate of Calibration.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty