Would you like to log in again?
Universal 10-32 PEEK Column Connectors and Plugs, Restek
Supplier: Restek
25016 25015
RK25016PK 85.09 USD InStock
RK25016 RK25015
Universal 10-32 PEEK Column Connectors and Plugs, Restek
Fittings and Connectors
Universal PEEK connectors allow easy installation of all 1/16" tubing, including stainless steel, up to 5,000 psi.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty