Would you like to log in again?
Universal Crimper Press, PerkinElmer
Supplier: PerkinElmer
B0139005
10137-514EA 6614.67 USD InStock
10137-514
Universal Crimper Press, PerkinElmer
Crimpers, Cappers and Decappers
Universal crimper press provides high-quality pan sealing for the autosampler sample pans/hermetically sealed pans.
  • Crimp standard aluminum pans, large-volume stainless steel (24 bar), and volatile pans
  • Includes standard Sample Pan Manipulator
  • Provided with inserts to seal various autosampler sample pans
The Universal Crimper Press can crimp the standard aluminium pans, O-Ring sealed large-volume stainless steel pans, and the 20 mL volatile sample pans.
Delivery information: Includes sample pan suction tool and inserts to seal 10, 30 and 50 µL sample pans.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty