Would you like to log in again?
FGV Digital Force Gauge, SEALS USA INC
Supplier: SEALS USA INC
FGV-100XY FGV-10XY FGV-5XY FGV-0.5XY FGV-1XY FGV-2XY FGV-200XY FGV-20XY FGV-50XY
97024-764EA 1789.25 USD InStock
97024-764 97024-754 97024-766 97024-756 97024-760 97024-762 97024-752 97024-768 97024-758
FGV Digital Force Gauge, SEALS USA INC
Dynamometers
The Series FGV-XY Force Gauges combine a rugged, ergonomic and compact design to provide a complete force testing instrument
On-board statistical calculations provide fast access to maximum, minimum, average and standard deviation data. Units include the free TORIEMON-USB software and USB cable allowing further data analysis on a PC. The 1,000 point memory allows for data exporting in real time or after testing is complete. In addition, the RS-232C and analog outputs provide the FGV with maximum communication flexibility. Models are available from 0.5 lbs to 200 lb ranges providing a large offering for various testing requirements.
The 180° reversible display combined with the adjustable force direction and dual labeled keypad enable the FGV-XY to excel in both compression and tension applications in any direction. Typical applications include wallboard, wire pull, wire crimps, trigger pull, brake pedal, aerospace ceramic, springs, food ripeness, corrugated paper strength, blister pack, medical component, muscle strength, ergonomic testing, adhesives, syringe plunger, needle sharpness and many more.
These instruments are utilized in a variety of applications such as, incoming quality inspection, finished goods testing, R&D or almost any portable or force stand testing requirement.
Certifications: CE Compliant.
Ordering information: Carrying Case Included, AC Adapter/Charger Included, Hook, Chisel, Flat Head, Notched Head, Hanger, Cone Head, Thread Adapter, Extension Rod, USB Cable all Included, 2-Year Warranty, NIST Certification Optional.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty