Would you like to log in again?
AlbumiNZ™ Bovine Serum Albumin (BSA)
Supplier: MP Biomedicals
AlbumiNZ
0219989980 0219989925 0219989910 0219989990 0219989950 0219989991
IC219989910EA 146.63 USD InStock
IC219989910 10273-264 IC219989990 IC219989980 IC219989991 IC219989925
AlbumiNZ™ Bovine Serum Albumin (BSA)
Proteins and Peptides
AlbumiNZ™ Low Fatty Acid BSA is chromatographically purified from New Zealand-sourced bovine plasma in an ISO quality system assuring complete traceability and consistent high quality. This albumin has very low endotoxin and low free fatty acids, is tested for the absence of viruses according to 9CFR and has the highest possible biosafety level.
  • Stem Cell Grade
  • Low Free Fatty Acid
  • Presentation: Off-white Powder
  • Moisture: ≤5%
AlbumiNZ™ Low Fatty Acid BSA supports growth of stem cells and increases protein production from hybridoma cells.
Certain conformational and primary-sequence epitopes of BSA are suspected allergens in human beef and milk allergies.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty