Would you like to log in again?
Cotton Swabs
Supplier: Techspray
2301-100
14220-263CS 537.79 USD InStock
14220-263
Cotton Swabs
Swabs Swabs for Cleaning
Absorbent cotton swab is tightly wrapped with precleaned cotton ensures the least amount of extractable contaminants.
  • Solvent resistant
  • Low extractables
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty