Would you like to log in again?
Fat Mat® Oil-Only Absorbent Mat Pad in Dispenser Box, PIG®
Supplier: New Pig
MAT4101
10860-052BX 174.5 USD InStock
10860-052
Fat Mat® Oil-Only Absorbent Mat Pad in Dispenser Box, PIG®
Spill Control Materials
16 layers of 100% polypropylene are thermally bonded to form one highly absorbent mat capable of drinking up huge volumes of oily liquids.
  • Absorbs and retains oils and fuels
  • Keep moisture away from surface
  • Won't rip, tear, fray or create a trip hazard
Double-thick version of PIG Oil-Only Mat has 16 layers of 100% polypropylene, which are thermally bonded to form one highly absorbent mat capable of drinking up huge volumes of oily liquids. Tough, spunbond top layer stands up to heavy foot traffic; won't rip, tear, fray or create a trip hazard, even when saturated. Fast-wicking "diaper action" transfers oil into the absorbent inner layers to keep moisture away from surface. Absorbs and retains oils and fuels, not water.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty