Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
PUF Glass Holders, Restek
Supplier: Restek
22965 22964
10863-058EA 50.6 USD InStock
10863-058 10862-926
PUF Glass Holders, Restek
Samplers Air Samplers
Durable and reusable, PUF glass holders reduce waste and a cost-effective alternative to precleaned packed cartridges.
  • Fit either 6 cm or 22 mm O-Ø PUF plug or can be used with bulk SDVB resin
  • Large glass holder fitted with double stainless steel screens for support
  • Small glass holder has stem designed for secure 1/4" I-Ø tubing connection to sampling pump
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty