Would you like to log in again?
Air Canister Tripod, Restek
Supplier: Restek
24151
10863-240EA 534.94 USD InStock
10863-240
Air Canister Tripod, Restek
Tripods
Restek’s air canister tripod holds two canisters simultaneously for collocated ambient air sampling.
  • Lightweight (12 lbs.) and compact for easy storage and transport
  • Extends from 6 to 9' high
  • Large base provides enhanced stability without additional supports
  • Sturdy, rugged metal design for outdoor sampling and transport
The custom-designed bracket holds most 1, 3, and 6 L canisters securely without any tools.
Ordering information: Air sampling canisters sold separately.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty