Would you like to log in again?
Buchi® Vacuum Pump, V-100
Supplier: Büchi
Buchi®
11V100010 11V100111 11V100000 11V100110
10746-764EA 4246.03 USD InStock
10746-764 10746-758 10746-756 10746-770
Buchi® Vacuum Pump, V-100
Pumps Vacuum Pumps
The Vacuum Pump V-100 is an economical and chemically resistant PTFE diaphragm vacuum pump with a space saving design
  • On/off-regulated by the optional Interface I-100
  • Optimally suited to work with the Rotavapor R-100
It covers the essential needs in vacuum generation.
Ordering information: Pump available with or without interface, secondary condenser, or Woulff bottle. Additional accessories are required for adaptation with older BUCHI systems. Please contact your VWR representative for further assistance.
Delivery information: The ready to use vacuum pump will be delivered with a silencer and 2 m of vacuum tubing.
Order Now
ORDER
ACCESSORIES
SERVICES
Services: BUCHI provides convenient PM Mail - in services. After ordering, you will receive a protective box to ship your equipment to BUCHI Corporation along with a packaging instruction. Upon receipt, your equipment will be inspected for any defects and the performance of your instrument will be compared to manufacturer standards. After the PM, a service report and your equipment will be carefully repacked then returned to you with standard ground shipping.
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty