Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Nε-carbobenzoxy-L-lysine

Supplier: Matrix Scientific
Synonyms: H-Lys(Z)-OH(S)-2-Amino-6-(((benzyloxy)carbonyl)amino)hexanoic acidNε-Cbz-L-lysine

065138-100G
102551-664EA 290.77 USD InStock
102551-664
Nε-carbobenzoxy-L-lysine
Nepsilon-carbobenzoxy-L-lysine
CAS: 1155-64-2
MDL Number: MFCD00002638
Molecular Formula: C14H20N2O4
Molecular Weight: 280,33
Hazard: Irritant


Formula: C₁₄H₂₀N₂O₄
MW: 280.32 g/mol
Melting Pt: 259 °C (dec.)
MDL Number: MFCD00002638
CAS Number: 1155-64-2
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty