Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Amino-4-methyl-5-pyrimidineboronic acid pinacol ester ≥97%

Supplier: Matrix Scientific
Synonyms: 4-Methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrimidin-2-amine4-Methyl-2-aminopyrimidine-5-boronic acid, pinacol ester

071106-1G 071106-5G
102519-484EA 1969.23 USD InStock
102519-484 102519-482
2-Amino-4-methyl-5-pyrimidineboronic acid pinacol ester ≥97%
2-Amino-4-methyl-5-pyrimidineboronic acid pinacol ester
Hazard: Irritant


Formula: C₁₁H₁₈BN₃O₂
MW: 235.09 g/mol
MDL Number: MFCD18072554
CAS Number: 944401-55-2
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty