Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Bromo-5-nitrobenzaldehyde ≥95%

Supplier: Matrix Scientific

072427-1G 072427-5G
102524-786EA 627.69 USD InStock
102524-786 102524-784
3-Bromo-5-nitrobenzaldehyde ≥95%
3-Bromo-5-nitrobenzaldehyde
Hazard: Irritant


Formula: C₇H₄BrNO₃
MDL Number: MFCD00100097
CAS Number: 355134-13-3
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty