Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

5-Fluorosalicylaldehyde ≥95%

Supplier: Matrix Scientific
Synonyms: 5-Fluoro-2-hydroxybenzaldehyde

072525-5G
102525-312EA 27.69 USD InStock
102525-312
5-Fluorosalicylaldehyde ≥95%
5-Fluorosalicylaldehyde
Hazard: Irritant


Formula: C₇H₅FO₂
MW: 140.11 g/mol
MDL Number: MFCD01090997
CAS Number: 347-54-6
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty