Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

5-Amino-4,6-dichloropyrimidine ≥95%

Supplier: Matrix Scientific
Synonyms: 4,6-Dichloropyrimidin-5-ylamine

072885-25G
102551-814EA 146.15 USD InStock
102551-814
5-Amino-4,6-dichloropyrimidine ≥95%
5-Amino-4,6-dichloropyrimidine
Hazard: Irritant/Moisture Sensitive


Formula: C₄H₃Cl₂N₃
Melting Pt: 143…146 °C
MDL Number: MFCD00006108
CAS Number: 5413-85-4
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty