Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Amino-2-methylbenzoic acid ≥95%

Supplier: Matrix Scientific
Synonyms: 4-Amino-o-toluic acid

073080-5G
102528-288EA 309.23 USD InStock
102528-288
4-Amino-2-methylbenzoic acid ≥95%
4-Amino-2-methylbenzoic acid
Hazard: Irritant


Formula: C₈H₉NO₂
MW: 151.16 g/mol
MDL Number: MFCD06656133
CAS Number: 2486-75-1
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty