Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1,10-Phenanthroline ≥95%

Supplier: Matrix Scientific

073090-100G
102528-328EA 127.69 USD InStock
102528-328
1,10-Phenanthroline ≥95%
1,10-Phenanthroline
Hazard: Toxic


Formula: C₁₂H₈N₂
MW: 180.21 g/mol
Melting Pt: 97…102 °C
MDL Number: MFCD00011678
CAS Number: 66-71-7
UN: 2811
ADR: 6.1,III
https://us-legacy.vwr.com/stibo/web/std.lang.all/19/82/2351982.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty