Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Vanillic acid ≥95%

Supplier: Matrix Scientific
Synonyms: 4-Hydroxy-3-methoxybenzoic acid

073998-100G
102534-104EA 95.38 USD InStock
102534-104
Vanillic acid ≥95%
Vanillic acid
Hazard: Irritant


Formula: C₈H₈O₄
MW: 168.15 g/mol
MDL Number: MFCD00002551
CAS Number: 121-34-6
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty