Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Amino-3-hydroxybenzoic acid ≥95%

Supplier: Matrix Scientific

074144-5G 074144-100G 074144-10G
102534-880EA 769.23 USD InStock
102534-880 102534-876 102534-878
4-Amino-3-hydroxybenzoic acid ≥95%
4-Amino-3-hydroxybenzoic acid
Hazard: Irritant


Formula: C₇H₇NO₃
MW: 153.14 g/mol
MDL Number: MFCD00017094
CAS Number: 2374-03-0
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty