Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

5-Chloromethyl-1H-tetrazole ≥95%

Supplier: Matrix Scientific

074299-5G
102535-700EA 461.54 USD InStock
102535-700
5-Chloromethyl-1H-tetrazole ≥95%
5-Chloromethyl-1H-tetrazole
Hazard: Irritant


Formula: C₂H₃ClN₄
MDL Number: MFCD00512845
CAS Number: 55408-11-2
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty