Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

6-Chloro-m-toluidine ≥95%

Supplier: Matrix Scientific

074483-25G
102833-696EA 153.85 USD InStock
102833-696
6-Chloro-m-toluidine ≥95%
6-Chloro-m-toluidine
Hazard: Toxic


Formula: C₇H₈ClN
MW: 141.6 g/mol
Boiling Pt: 228…230 °C
Melting Pt: 29…30 °C
Flash Pt: 107 °C
MDL Number: MFCD00007674
CAS Number: 95-81-8
UN: 2239
ADR: 6.1,III
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty