Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Ethynylpyridine ≥95%

Supplier: Matrix Scientific

074697-5G
102537-756EA 60 USD InStock
102537-756
2-Ethynylpyridine ≥95%
2-Ethynylpyridine
Hazard: Irritant/Refrigerate


Formula: C₇H₅N
MW: 103.12 g/mol
Boiling Pt: 77…78 °C (14 mmHg)
Density: 1.02
Flash Pt: 76 °C
MDL Number: MFCD00041598
CAS Number: 1945-84-2
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty