Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(R)-(+)-α-Methylbenzylamine ≥95%

Supplier: Matrix Scientific
Synonyms: D(+)-α-Methylbenzylamine(R)-(+)-1-Phenylethylamine(R)-(+)-α-Phenylethylamine

074833-500G 074833-100G
102538-462EA 60 USD InStock
102538-462 102664-016
(R)-(+)-α-Methylbenzylamine ≥95%
(R)-(+)-α-Methylbenzylamine
Hazard: Corrosive/Air Sensitive


Formula: C₈H₁₁N
MW: 121.18 g/mol
Boiling Pt: 187 °C
Density: 0.948
Flash Pt: 71 °C
MDL Number: MFCD00064405
CAS Number: 3886-69-9
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty