Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

β-Alanine ≥95%

Supplier: Matrix Scientific
Synonyms: 3-Aminopropanoic acidH-β-Ala-OHH-Bala-OH3-Aminopropionic acid

074842-100G
102538-498EA 178.46 USD InStock
102538-498
β-Alanine ≥95%
β-Alanine
Hazard: Irritant


Formula: C₃H₇NO₂
MW: 89.09 g/mol
MDL Number: MFCD00008200
CAS Number: 107-95-9
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty