Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(4-(tert-butoxy)phenyl)propanoic acid ≥95%

Supplier: Matrix Scientific
Synonyms: O-tert-Butyl-Nalpha-Fmoc-D-tyrosineFmoc-D-Tyr(tBu)-OHN-Fmoc-O-tert-butyl-D-tyrosineNalpha-Fmoc-O-tert-butyl-D-tyrosine

074846-5G
102538-514EA 238.46 USD InStock
102538-514
(R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(4-(tert-butoxy)phenyl)propanoic acid ≥95%
(R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(4-(tert-butoxy)phenyl)propanoic acid
Hazard: Irritant


Formula: C₂₈H₂₉NO₅
MW: 459.5 g/mol
MDL Number: MFCD00065684
CAS Number: 118488-18-9
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty