Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Oxoglutaric acid ≥95%

Supplier: Matrix Scientific
Synonyms: 2-Ketoglutaric acid2-Oxopentanedioic acidA-Ketoglutaric acidα-Ketoglutaric acid

074886-100G
102538-696EA 60 USD InStock
102538-696
2-Oxoglutaric acid ≥95%
2-Oxoglutaric acid
Hazard: Irritant/Refrigerate


Formula: HOOCCH₂CH₂COCOOH
MW: 146.1 g/mol
Melting Pt: 114…117 °C
MDL Number: MFCD00004165
CAS Number: 328-50-7
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty