Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N-Boc-L-prolinol ≥95%

Supplier: Matrix Scientific
Synonyms: (S)-1-Boc-2-pyrrolidinemethanol(S)-1-(tert-Butoxycarbonyl)-2-pyrrolidinemethanolN-Boc-L-ProlinolN-(tert-Butoxycarbonyl)-L-prolinol

074946-100G
102538-988EA 230.77 USD InStock
102538-988
N-Boc-L-prolinol ≥95%
N-Boc-L-prolinol
Hazard: Irritant


Formula: C₁₀H₁₉NO₃
MW: 201.27 g/mol
MDL Number: MFCD00066232
CAS Number: 69610-40-8
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty