Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Bromoacetaldehyde dimethyl acetal ≥95%

Supplier: Matrix Scientific
Synonyms: 2-Bromo-1,1-dimethoxyethane

074951-100G
102539-012EA 78.46 USD InStock
102539-012
Bromoacetaldehyde dimethyl acetal ≥95%
Bromoacetaldehyde dimethyl acetal
Hazard: Flammable/Toxic


Formula: BrH₂CCH(OCH₃)₂
MW: 169.02 g/mol
Boiling Pt: 150 °C
Density: 1.435
MDL Number: MFCD00000213
CAS Number: 7252-83-7
UN: 2929
ADR: 6.1,II
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty