Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Aminothiazole ≥95%

Supplier: Matrix Scientific
Synonyms: Thiazol-2-ylamineThiazol-2-amine

074968-100G
102539-100EA 58.46 USD InStock
102539-100
2-Aminothiazole ≥95%
2-Aminothiazole
Hazard: Irritant


Formula: C₃H₄N₂S
MW: 100.14 g/mol
MDL Number: MFCD00005325
CAS Number: 96-50-4
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty