Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Methyl-2-amino-3-hydroxypropanoate hydrochloride ≥95%

Supplier: Matrix Scientific
Synonyms: H-DL-Ser-OMe·HCl3-hydroxy-1-methoxy-1-oxopropan-2-aminium chloridemethyl 2-amino-3-hydroxypropanoateMethyl DL-serinate hydrochlorideDL-Serine Methyl Ester HydrochlorideH-DL-Ser-OMe.HCl

074977-100G
102539-140EA 113.85 USD InStock
102539-140
Methyl-2-amino-3-hydroxypropanoate hydrochloride ≥95%
Methyl-2-amino-3-hydroxypropanoate hydrochloride
Hazard: Irritant/Moisture Sensitive


Formula: C₄H₁₀ClNO₃
MW: 155.578 g/mol
Melting Pt: 133…134 °C
MDL Number: MFCD00012593
CAS Number: 5619-04-5
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty