Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

L-Norvaline ≥95%

Supplier: Matrix Scientific
Synonyms: H-Nva-OHL(+)-Norvaline(S)-2-Aminopentanoic acid(S)-(+)-Norvaline

074980-100G
102539-156EA 276.92 USD InStock
102539-156
L-Norvaline ≥95%
L-Norvaline
Hazard: Irritant


Formula: C₅H₁₁NO₂
MW: 117.15 g/mol
Melting Pt: 307 °C
MDL Number: MFCD00064421
CAS Number: 6600-40-4
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty