Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

5-Nitrothiophene-2-carboxaldehyde ≥95%

Supplier: Matrix Scientific

074982-10G 074982-100G
102539-164EA 40 USD InStock
102539-164 102539-166
5-Nitrothiophene-2-carboxaldehyde ≥95%
5-Nitrothiophene-2-carboxaldehyde
Hazard: Irritant/Air Sensitive


Formula: C₅H₃NO₃S
MW: 157.15 g/mol
Melting Pt: 72…76 °C
MDL Number: MFCD00005433
CAS Number: 4521-33-9
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty